J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Phosphate
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Phosphate
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Phosphate
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Phosphate
This BAKER ANALYZED® Reagent is a phosphate buffer solution for laboratory, research, and/or manufacturing uses.
  • Contains no antimicrobial preservative
Each lot is compared to NIST Standard Reference Buffer.
Product Family Options
  • Reference Temperature
    Type de conditionnement
    Taille
    pH
    Color
  • 25 °C
    PE bottle
    500 ml
    7.00
    Colorless
    Each (500ml)
    Each (500ml) - $117.22
    $117.22
    Case of 12 (500ml)
    Case of 12 (500ml) - $1,080.11
    $1,080.11
    Each (500ml)
       $117.22
    Case of 12 (500ml)
       $1,080.11
     : BAKR5608-01
  • 25 °C
    Cubetainer®
    4000 ml
    7.00
    Colorless
    Each (4l)
    Each (4l) - $398.68
    $398.68
    Case of 4 (4l)
    Case of 4 (4l) - $1,352.97
    $1,352.97
    Each (4l)
       $398.68
    Case of 4 (4l)
       $1,352.97
     : BAKR5608-03