The J.T.Baker® BAKERBOND® Poly Hi-Propyl mixed mode hydrophobic interaction chromatography (HIC) resin provides improved selectivity for superior separation in process chromatography applications.
- Store between 2 and 15 °C
Powered by a proprietary surface chemistry combining hydrophobic functionality with weak anionic exchange sites, BAKERBOND® Poly Hi-Propyl mixed mode hydrophobic interaction chromatography media utilizes its unique selectivity to enable higher purification performance across a wide range of biopharmaceuticals.
Designed and manufactured by Avantor to the high standards established for our J.T.Baker® brand chemicals, BAKERBOND® Poly Hi-Propyl ion exchange resin can be used with other J.T.Baker® BAKERBOND® resins to enhance the separation power of your chromatography process steps.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable: