Cubic, Cubic Bodied and Cubic Faced Models
Catalogue # 470311-024
Fournisseur: Klinger
Spécifications
- Description:Cubic, Cubic Bodied and Cubic Faced Models
- Cat. no.:470311-024
Spécifications
À propos de cet article
These models are hand made in the USA by Klinger educational products.
- Bravais type lattice
- Crystal models
- Clearly labeled
These are permanent structures. We only use grade A materials. The 1" balls are made of hard maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.