Snap Caps, Sun Sri
Supplier: Thermo Scientific
Certificates
About this item
These snap caps eliminate crimping and decrimping of closures with easy to apply snap caps and also it provides a secure seal, minimizing evaporation even with volatile samples.
- Septa pre-assembled
- Compatible with StepVials and other snap-ring style vials
- Not compatible with a standard 11 mm aluminum seal finish
The audible click ensures that a secure seal has been formed and the cap is properly applied.
Frequently Bought Together