Babcock Cream Test Bottle Brush, Justman
Catalog # 30620-540
Supplier: Justman Brush
Specifications
- Description:Babcock Cream Bottle Brush
- Total length:28 cm (11")
- Cat. no.:30620-540
- Supplier No.:1707
Specifications
About this item
Babcock cream bottle brush with fan tip.