First Strand Buffer, 5X, Teknova
First Strand Buffer, 5X, Teknova
First Strand Buffer, 5X, 500 ml
Catalog # CA100217-306
Supplier:   Teknova
Supplier #:  F1204
First Strand Buffer, 5X, Teknova
Catalog # CA100217-306
Supplier:  Teknova
CAS Number:  
First Strand Buffer, 5X, Teknova
First Strand Buffer, 5X, 500 ml
CAS Number:  
Price
$811.79
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
First strand synthesis buffer designed for use in reverse transcription.
Consists of Tris-HCl (250 mM), pH 8.3, potassium chloride (375 mM), magnesium chloride (15 mM) and DTT (50 mM).
Specifications
Specifications
  • Désignation:
    First Strand Buffer, 5X
  • Taille:
    500 ml