DYMO Compatible Cryogenic Labels
DYMO Compatible Cryogenic Labels
Certificates
DYMO Compatible Cryogenic Labels
DYMO Compatible Cryogenic Labels
68 Options Available Below
Product Details & Documents
DYMO compatible direct thermal cryo labels with permanent adhesive.
  • Temperature range: –196 to +70 °C (–321 to +158 °F)
  • Compatible with all DYMO label writers (excluding models 550, 550Turbo, and 5XL)
  • Resists extreme cryogenic temperatures (–196° C in liquid phase and vapour phase liquid nitrogen)
  • Ideal for the identification of 0.5 to 5 ml vials and tubes
DYMO compatible cryo labels are available in a range of sizes, shapes, and colors, perfect for identifying 0.5 to 5 ml cryo vials and microtubes, vial caps/tops, cryo/freezer boxes, as well as microplates, petri dishes, and cell culture plates.
These cryogenic labels are designed for short and long-term cryo-preservation in ultra-low temperature freezers (–80 °C/–112 °F), liquid phase and vapor phase liquid nitrogen (–196 °C/–321 °F), and transportation on dry ice. They will also withstand temperatures up to 70 °C (158 °F), ideal for various applications throughout the lab, including use in incubators.
Product Family Options
Circle permanent DYMO compatible cryogenic labels
Product Information
  • Désignation
    Color
    Core Size
    Dimensions
    Packaging
    Availability
    Price
  • DYMO compatible cryogenic label
    White
    1"
    9 mm (0.35")
    2000 labels/roll (2 labels across)
    Discontinued
    Supplier Number: EF1F-030WH
  • DYMO compatible cryogenic label
    Yellow
    1"
    9 mm (0.35")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $168.85
    $168.85
    Roll of 1
       $168.85
    Supplier Number: 807656024523
  • DYMO compatible cryogenic label
    Blue
    1"
    9 mm (0.35")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $168.85
    $168.85
    Roll of 1
       $168.85
    Supplier Number: 807656024530
  • DYMO compatible cryogenic label
    Red
    1"
    9 mm (0.35")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $166.78
    $166.78
    Roll of 1
       $166.78
    Supplier Number: 807656024547
  • DYMO compatible cryogenic label
    Bright green
    1"
    9 mm (0.35")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $166.78
    $166.78
    Roll of 1
       $166.78
    Supplier Number: 807656024554
  • DYMO compatible cryogenic label
    Orange
    1"
    9 mm (0.35")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $168.40
    $168.40
    Roll of 1
       $168.40
    Supplier Number: 807656024561
  • DYMO compatible cryogenic label
    White
    1"
    12.7 mm (0.5")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $165.97
    $165.97
    Roll of 1
       $165.97
    Supplier Number: 807656005324
  • DYMO compatible cryogenic label
    Yellow
    1"
    12.7 mm (0.5")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $208.46
    $208.46
    Roll of 1
       $208.46
    Supplier Number: 807656006642
  • DYMO compatible cryogenic label
    Blue
    1"
    12.7 mm (0.5")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $208.46
    $208.46
    Roll of 1
       $208.46
    Supplier Number: 807656006635
  • DYMO compatible cryogenic label
    Red
    1"
    12.7 mm (0.5")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $208.46
    $208.46
    Roll of 1
       $208.46
    Supplier Number: 807656006628
  • DYMO compatible cryogenic label
    Bright green
    1"
    12.7 mm (0.5")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $208.46
    $208.46
    Roll of 1
       $208.46
    Supplier Number: 807656006611
  • DYMO compatible cryogenic label
    Orange
    1"
    12.7 mm (0.5")
    2000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $208.46
    $208.46
    Roll of 1
       $208.46
    Supplier Number: 807656006277
Rectangle permanent DYMO compatible cryogenic labels
Product Information
  • Désignation
    Color
    Core Size
    Dimensions
    Packaging
    Availability
    Price
  • DYMO compatible cryogenic label
    White
    1"
    57×32 mm (2.25×1.25")
    1000 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1,000
    Roll of 1,000 - $221.48
    $221.48
    Roll of 1,000
       $221.48
    Supplier Number: 807656032900
  • DYMO compatible cryogenic label
    Red
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $107.03
    $107.03
    Roll of 1
       $107.03
    Supplier Number: 807656024219
  • DYMO compatible cryogenic label
    Blue
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $108.07
    $108.07
    Roll of 1
       $108.07
    Supplier Number: 807656024196
  • DYMO compatible cryogenic label
    Bright green
    1"
    12.7×25.4 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $108.07
    $108.07
    Roll of 1
       $108.07
    Supplier Number: 807656024202
  • DYMO compatible cryogenic label
    White
    1"
    13×25 mm (0.5×1")
    1000 labels/roll (2 labels across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $82.23
    $82.23
    Roll of 1
       $82.23
    Supplier Number: 807656005355
  • DYMO compatible cryogenic label
    White
    1"
    14×87 mm (0.56×3.44")
    350 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $85.41
    $85.41
    Roll of 1
       $85.41
    Supplier Number: 807656006864
  • DYMO compatible cryogenic label
    White
    1"
    18×38 mm (0.7×1.49")
    500 labels/roll (1 label across)
    Discontinued
    Supplier Number: 807656005898
  • DYMO compatible cryogenic label
    Red
    1"
    18×38 mm (0.7×1.49")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $94.10
    $94.10
    Roll of 1
       $94.10
    Supplier Number: 807656024226
  • DYMO compatible cryogenic label
    White
    1"
    19×38.1 mm (0.75×1.5")
    750 labels/roll (1 label across)
    Discontinued
    Supplier Number: 807656024233
  • DYMO compatible cryogenic label
    White
    1"
    20×45 mm (0.79×1.77")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $74.57
    $74.57
    Roll of 1
       $74.57
    Supplier Number: 807656024240
  • DYMO compatible cryogenic label
    White
    1"
    25.4×25.4 mm (1×1")
    750 labels/roll (1 label across)
    Discontinued
    Supplier Number: 807656006512
  • DYMO compatible cryogenic label
    Grey
    1"
    25.4×25.4 mm (1×1")
    750 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $139.09
    $139.09
    Roll of 1
       $139.09
    Supplier Number: 807656024257
  • DYMO compatible cryogenic label
    White
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Discontinued
    Supplier Number: 807656005232
  • DYMO compatible cryogenic label
    Yellow
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $102.99
    $102.99
    Roll of 1
       $102.99
    Supplier Number: 807656024264
  • DYMO compatible cryogenic label
    Pink
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $102.99
    $102.99
    Roll of 1
       $102.99
    Supplier Number: 807656024271
  • DYMO compatible cryogenic label
    Blue
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $104.02
    $104.02
    Roll of 1
       $104.02
    Supplier Number: 807656005461
  • DYMO compatible cryogenic label
    Red
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $102.99
    $102.99
    Roll of 1
       $102.99
    Supplier Number: 807656024288
  • DYMO compatible cryogenic label
    Orange
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $102.99
    $102.99
    Roll of 1
       $102.99
    Supplier Number: 807656024295
  • DYMO compatible cryogenic label
    Bright green
    1"
    25.4×54 mm (1×2.13")
    500 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $104.02
    $104.02
    Roll of 1
       $104.02
    Supplier Number: 807656024301
  • DYMO compatible cryogenic label
    Blue
    1"
    29×89 mm (1.13×3.5")
    350 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $105.97
    $105.97
    Roll of 1
       $105.97
    Supplier Number: 807656024332
  • DYMO compatible cryogenic label
    Red
    1"
    29×89 mm (1.13×3.5")
    350 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $105.97
    $105.97
    Roll of 1
       $105.97
    Supplier Number: 807656024349
  • DYMO compatible cryogenic label
    Bright green
    1"
    29×89 mm (1.13×3.5")
    350 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $105.97
    $105.97
    Roll of 1
       $105.97
    Supplier Number: 807656024356
  • DYMO compatible cryogenic label
    Orange
    1"
    29×89 mm (1.13×3.5")
    350 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $105.97
    $105.97
    Roll of 1
       $105.97
    Supplier Number: 807656024363
  • DYMO compatible cryogenic label
    White
    1"
    29×89 mm (1.13×3.5")
    350 labels/roll (1 label across)
    Discontinued
    Supplier Number: 807656005348
  • DYMO compatible cryogenic label
    Yellow
    1"
    29×89 mm (1.13×3.5")
    350 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $105.97
    $105.97
    Roll of 1
       $105.97
    Supplier Number: 807656024325
  • DYMO compatible cryogenic label
    White
    1"
    50.8×6.35 mm (2×0.25")
    1000 labels/roll (3 stacked labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $91.17
    $91.17
    Roll of 1
       $91.17
    Supplier Number: 807656024370
  • DYMO compatible cryogenic label
    Yellow
    1"
    50.8×6.35 mm (2×0.25")
    1000 labels/roll (3 stacked labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $112.80
    $112.80
    Roll of 1
       $112.80
    Supplier Number: 807656024387
  • DYMO compatible cryogenic label
    Blue
    1"
    50.8×6.35 mm (2×0.25")
    1000 labels/roll (3 stacked labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $112.80
    $112.80
    Roll of 1
       $112.80
    Supplier Number: 807656024394
  • DYMO compatible cryogenic label
    Bright green
    1"
    50.8×6.35 mm (2×0.25")
    1000 labels/roll (3 stacked labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $112.80
    $112.80
    Roll of 1
       $112.80
    Supplier Number: 807656024400
  • DYMO compatible cryogenic label
    Red
    1"
    50.8×6.35 mm (2×0.25")
    1000 labels/roll (3 stacked labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $112.80
    $112.80
    Roll of 1
       $112.80
    Supplier Number: 807656024417
  • DYMO compatible cryogenic label
    Orange
    1"
    50.8×6.35 mm (2×0.25")
    1000 labels/roll (3 stacked labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $112.80
    $112.80
    Roll of 1
       $112.80
    Supplier Number: 807656024424
  • DYMO compatible cryogenic label
    White
    1"
    54×101 mm (2.13×3.9")
    300 labels/roll (1 label across)
    Log in for availability
    Log in for availability
    Roll of 3,000
    Roll of 3,000 - $135.21
    $135.21
    Roll of 3,000
       $135.21
    Supplier Number: 807656032917
Rectangle and circle permanent DYMO compatible cryogenic labels
Product Information
  • Désignation
    Color
    Core Size
    Dimensions
    Packaging
    Availability
    Price
  • DYMO compatible cryogenic label
    Beige
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032788
  • DYMO compatible cryogenic label
    Blue
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032771
  • DYMO compatible cryogenic label
    Orange
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032726
  • DYMO compatible cryogenic label
    Red
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032696
  • DYMO compatible cryogenic label
    Violet
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032719
  • DYMO compatible cryogenic label
    Yellow
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032702
  • DYMO compatible cryogenic label
    Dark brown
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032764
  • DYMO compatible cryogenic label
    Bright green
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.85
    $182.85
    Roll of 500
       $182.85
    Supplier Number: 807656032757
  • DYMO compatible cryogenic label
    Grey
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.85
    $182.85
    Roll of 500
       $182.85
    Supplier Number: 807656032740
  • DYMO compatible cryogenic label
    Yellow
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032733
  • DYMO compatible cryogenic label
    White
    1"
    12.7×26.4+9.5 mm (0.5×1.04+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $145.81
    $145.81
    Roll of 500
       $145.81
    Supplier Number: 807656032689
  • DYMO compatible cryogenic label
    White
    1"
    20×5.1+6.35 mm (0.79×0.2+0.25")
    1000 labels/roll (3 sets: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $116.44
    $116.44
    Roll of 1
       $116.44
    Supplier Number: 807656024431
  • DYMO compatible cryogenic label
    Blue
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $194.13
    $194.13
    Roll of 1
       $194.13
    Supplier Number: 807656024455
  • DYMO compatible cryogenic label
    Yellow
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $194.13
    $194.13
    Roll of 1
       $194.13
    Supplier Number: 807656024462
  • DYMO compatible cryogenic label
    Bright green
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $194.13
    $194.13
    Roll of 1
       $194.13
    Supplier Number: 807656024479
  • DYMO compatible cryogenic label
    Orange
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $194.13
    $194.13
    Roll of 1
       $194.13
    Supplier Number: 807656024486
  • DYMO compatible cryogenic label
    Red
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $194.13
    $194.13
    Roll of 1
       $194.13
    Supplier Number: 807656024493
  • DYMO compatible cryogenic label
    Pink
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $194.13
    $194.13
    Roll of 1
       $194.13
    Supplier Number: 807656024509
  • DYMO compatible cryogenic label
    White
    1"
    23.9×12.7+11.1 mm (0.94×0.5+0.44")
    700 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 700
    Roll of 700 - $121.96
    $121.96
    Roll of 700
       $121.96
    Supplier Number: 807656032924
  • DYMO compatible cryogenic label
    Beige
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032818
  • DYMO compatible cryogenic label
    Blue
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032870
  • DYMO compatible cryogenic label
    Dark brown
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.85
    $182.85
    Roll of 500
       $182.85
    Supplier Number: 807656032856
  • DYMO compatible cryogenic label
    Red
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.85
    $182.85
    Roll of 500
       $182.85
    Supplier Number: 807656032849
  • DYMO compatible cryogenic label
    Violet
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.85
    $182.85
    Roll of 500
       $182.85
    Supplier Number: 807656032832
  • DYMO compatible cryogenic label
    Yellow
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032863
  • DYMO compatible cryogenic label
    White
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $145.81
    $145.81
    Roll of 500
       $145.81
    Supplier Number: 807656032795
  • DYMO compatible cryogenic label
    Bright green
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032887
  • DYMO compatible cryogenic label
    Grey
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.85
    $182.85
    Roll of 500
       $182.85
    Supplier Number: 807656032801
  • DYMO compatible cryogenic label
    Yellow
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $184.62
    $184.62
    Roll of 500
       $184.62
    Supplier Number: 807656032825
  • DYMO compatible cryogenic label
    Orange
    1"
    26.4×12.7+9.5 mm (1.04×0.5+0.38")
    500 labels/roll (1 set: rectangle+circle)
    Log in for availability
    Log in for availability
    Roll of 500
    Roll of 500 - $182.89
    $182.89
    Roll of 500
       $182.89
    Supplier Number: 807656032894