MOPS/SDS, 20X, Running Buffer, Teknova | Avantor
MOPS/SDS, 20X, Running Buffer, Teknova
Supplier:   Teknova
MOPS/SDS, 20X, Running Buffer, Teknova
Supplier:  Teknova
Certificates
MOPS/SDS, 20X, Running Buffer, Teknova
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
MOPS/SDS running buffer is designed for protein electrophoresis.
  • Sterile
20X liquid concentrate contains 1 M MOPS, 1 M Tris, 20.5 mM EDTA and 69.3 mM SDS.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • 20X MOPS/SDS Running Buffer
    4 L
    Discontinued
    Supplier Number: -
  • 20X MOPS/SDS Running Buffer
    500 mL
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500ml)
    Each (500ml) - $739.33
    $739.33
    Each (500ml)
       $739.33
    Supplier Number: M1089