Color Polyester Labels, Brady
Color Polyester Labels, Brady
Color Polyester Labels, Brady
Color Polyester Labels, Brady
Color Polyester Labels, Brady
Add color to reduce errors and increase efficiency with B-494 color polyester labels. The design allows users to differentiate lab samples in a snap.
  • Six color options that allow users to distinguish many different samples
  • Custom options to fit specific process and glassware needs
  • Efficient label creation by adding the Brady workstation laboratory identification software suite
Color printing below the label surface withstands the harshest lab environments, including liquid nitrogen, hot water baths, and more. Print is resistant to common lab chemicals like ethanol and xylene with the R6400 ribbon built into the cartridge.
Product Family Options
For Use with BradyPrinter i5100, BradyPrinter i7100, BRADY IP® Series Label Printer
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Dimensions
    Packaging
  • Colored polyester labels
    Polyester
    White/yellow
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $539.57
    $539.57
    Roll of 1
       $539.57
     : THT-250-494-YL
  • Colored polyester labels
    Polyester
    Red/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $539.57
    $539.57
    Roll of 1
       $539.57
     : THT-250-494-RD
  • Colored polyester labels
    Polyester
    Blue/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $539.57
    $539.57
    Roll of 1
       $539.57
     : THT-250-494-BL
  • Colored polyester labels
    Polyester
    Orange/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $539.57
    $539.57
    Roll of 1
       $539.57
     : THT-250-494-OR
  • Colored polyester labels
    Polyester
    Green/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $539.57
    $539.57
    Roll of 1
       $539.57
     : THT-250-494-GN
  • Colored polyester labels
    Polyester
    Pink/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $539.57
    $539.57
    Roll of 1
       $539.57
     : THT-250-494-PK
  • Polyester printer labels
    Polyester
    White/yellow
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $523.72
    $523.72
    Roll of 1
       $523.72
     : THT-179-494-YL
  • Polyester printer labels
    Polyester
    Red/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $507.99
    $507.99
    Roll of 1
       $507.99
     : THT-179-494-RD
  • Polyester printer labels
    Polyester
    Blue/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $507.99
    $507.99
    Roll of 1
       $507.99
     : THT-179-494-BL
  • Polyester printer labels
    Polyester
    Orange/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $507.99
    $507.99
    Roll of 1
       $507.99
     : THT-179-494-OR
  • Polyester printer labels
    Polyester
    Green/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $507.99
    $507.99
    Roll of 1
       $507.99
     : THT-179-494-GN
  • Polyester printer labels
    Polyester
    Pink/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $507.99
    $507.99
    Roll of 1
       $507.99
     : THT-179-494-PK
  • Colored polyester labels
    Polyester
    White/yellow
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $430.86
    $430.86
    Roll of 1
       $430.86
     : THT-152-494-YL
  • Colored polyester labels
    Polyester
    Red/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $430.86
    $430.86
    Roll of 1
       $430.86
     : THT-152-494-RD
  • Colored polyester labels
    Polyester
    Blue/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $430.86
    $430.86
    Roll of 1
       $430.86
     : THT-152-494-BL
  • Colored polyester labels
    Polyester
    Orange/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $430.86
    $430.86
    Roll of 1
       $430.86
     : THT-152-494-OR
  • Colored polyester labels
    Polyester
    Green/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $430.86
    $430.86
    Roll of 1
       $430.86
     : THT-152-494-GN
  • Colored polyester labels
    Polyester
    Pink/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    3000 labels/roll
    Roll of 1
    Roll of 1 - $430.86
    $430.86
    Roll of 1
       $430.86
     : THT-152-494-PK
For Use with BradyPrinter BBP®33 Label Printer, BradyPrinter i3300
  • Désignation
    Matière
    Color
    Pour l'utilisation avec
    Diameter
    Dimensions
    Packaging
  • Colored polyester labels
    White/yellow
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $264.83
    $264.83
    Roll of 1
       $264.83
     : B33-250-494-YL
  • Colored polyester labels
    Red/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $264.81
    $264.81
    Roll of 1
       $264.81
     : B33-250-494-RD
  • Colored polyester labels
    Blue/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $264.83
    $264.83
    Roll of 1
       $264.83
     : B33-250-494-BL
  • Colored polyester labels
    Orange/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $264.81
    $264.81
    Roll of 1
       $264.81
     : B33-250-494-OR
  • Colored polyester labels
    Green/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $264.83
    $264.83
    Roll of 1
       $264.83
     : B33-250-494-GN
  • Colored polyester labels
    Pink/white
    1.5 - 2 ml vials/tubes
    0.375" (Secondary Label Ø)
    12.7×25.4 mm (0.5×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $264.81
    $264.81
    Roll of 1
       $264.81
     : B33-250-494-PK
  • Polyester printer labels
    Polyester
    White/yellow
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $261.33
    $261.33
    Roll of 1
       $261.33
     : B33-179-494-YL
  • Polyester printer labels
    Polyester
    Red/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $261.33
    $261.33
    Roll of 1
       $261.33
     : B33-179-494-RD
  • Polyester printer labels
    Polyester
    Blue/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $261.33
    $261.33
    Roll of 1
       $261.33
     : B33-179-494-BL
  • Polyester printer labels
    Polyester
    Orange/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $250.23
    $250.23
    Roll of 1
       $250.23
     : B33-179-494-OR
  • Polyester printer labels
    Polyester
    Green/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $261.33
    $261.33
    Roll of 1
       $261.33
     : B33-179-494-GN
  • Polyester printer labels
    Polyester
    Pink/white
    2 - 8 ml vials/tubes
    25.4×25.4 mm (1×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $250.23
    $250.23
    Roll of 1
       $250.23
     : B33-179-494-PK
  • Colored polyester labels
    Polyester
    White/yellow
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $211.07
    $211.07
    Roll of 1
       $211.07
     : B33-152-494-YL
  • Colored polyester labels
    Polyester
    Red/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $211.07
    $211.07
    Roll of 1
       $211.07
     : B33-152-494-RD
  • Colored polyester labels
    Polyester
    Blue/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $171.03
    $171.03
    Roll of 1
       $171.03
     : 149808
  • Colored polyester labels
    Polyester
    Orange/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $211.07
    $211.07
    Roll of 1
       $211.07
     : B33-152-494-OR
  • Colored polyester labels
    Polyester
    Green/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $171.03
    $171.03
    Roll of 1
       $171.03
     : 149810
  • Colored polyester labels
    Polyester
    Pink/white
    1 - 8 ml vials/tubes
    9.5×25.4 mm (0.38×1")
    1500 labels/roll
    Roll of 1
    Roll of 1 - $211.07
    $211.07
    Roll of 1
       $211.07
     : B33-152-494-PK