ContactUsLinkComponent
Quick-RNA™ Plus Kits, Zymo Research
Quick-RNA™ Plus Kits, Zymo Research
Supplier:  Zymo Research
Certificates
undefined
Quick-RNA™ Plus Kits, Zymo Research
Supplier:  Zymo Research
Specifications
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Sample size:
    ≤1×107 Cells or 50 mg Tissue
  • Description:
    Quick-RNA™ Miniprep Plus Kit
  • Sample capacity:
    100 µg
  • Yield:
    spin column, 100 µg RNA (binding capacity), ≥50 µl (elution volume)