Pre-Printed Laboratory Warning Labels
À propos de cet article
Various pre-printed laboratory warning labels designed to advise staff and visitors of potential hazards and dangers. This includes biohazard and radioactive warning labels, as well as standardized THC warning labels for labeling a wide range of cannabis products.
- Temperature range: −80 to +100 °C (−112 to +212 °F)
- Withstands ultra-low temperature conditions (−80 °C/−112 °F)
- Special construction protects against abrasion, chemical exposure, and tearing
- Durable and high-quality materials for long-lasting use
Pre-printed general warning labels for identifying equipment, containers, and material that may pose a safety risk in the laboratory environment. With a wide temperature resistance range, these labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, etc.), hot surfaces, including water baths and heating blocks, as well as on equipment, instruments, and tools with moving parts.
13 Options Available Below
Product Details & Documents
Various pre-printed laboratory warning labels designed to advise staff and visitors of potential hazards and dangers. This includes biohazard and radioactive warning labels, as well as standardized THC warning labels for labeling a wide range of cannabis products.
- Temperature range: −80 to +100 °C (−112 to +212 °F)
- Withstands ultra-low temperature conditions (−80 °C/−112 °F)
- Special construction protects against abrasion, chemical exposure, and tearing
- Durable and high-quality materials for long-lasting use
Pre-printed general warning labels for identifying equipment, containers, and material that may pose a safety risk in the laboratory environment. With a wide temperature resistance range, these labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, etc.), hot surfaces, including water baths and heating blocks, as well as on equipment, instruments, and tools with moving parts.
Product Family Options
Informations sur le produit
- DésignationMatièreLegendColorCore SizeDimensionsPackagingAvailabilityPrice
- Warning labels - BiohazardPlastic−-−0.5"1000 labels/roll (1 label across)Discontinued-Supplier Number: -
- Warning labels - BiohazardPlasticBlack on red−1.125×1.125"1000 labels/roll (1 label across)Discontinued-Supplier Number: -
- Warning labels - BiohazardPlasticBlack on yellow−1.75×0.75"250 labels/roll (1 label across)Discontinued-Supplier Number: -
- Warning labels - RadioactivePlasticBlack on yellow3"12.7 mm (0.5″)1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 178,78 $Supplier Number: 807656026626
- Warning labels - Hot Surface HazardPlasticBlack/yellow on silver3"19.1×19.1 mm (0.75×0.75")250 labels/roll (1 label across)Log in pour la disponibilitéRoll of 191,93 $Supplier Number: 807656083971
- Warning labels - Hot Surface HazardPlasticBlack/yellow on silver3"19.1×19.1 mm (0.75×0.75")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1328,22 $Supplier Number: 807656026565
- Warning labels - Low TemperaturePlasticBlack/yellow on silver3"19.1×19.1 mm (0.75×0.75")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1328,22 $Supplier Number: 807656026602
- Warning labels - General Warning SymbolPlasticBlack/yellow on silver3"19.1×19.1 mm (0.75×0.75")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1328,22 $Supplier Number: 807656026589
- Warning labels - Low TemperaturePlasticBlack/yellow on silver3"19.1×19.1 mm (0.75×0.75")250 labels/roll (1 label across)Log in pour la disponibilitéRoll of 191,93 $Supplier Number: 807656026596
- Warning labels - HMISPlasticMulticolour3"22.2×22.2 mm (0.875×0.875")500 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1137,46 $Supplier Number: 807656084077
- Warning labels - HMISPlasticMulticolour3"25.4×76.2 mm (1×3")500 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1147,23 $Supplier Number: 807656026657
- Warning labels - NFPAPlasticMulticolour3"28.6×28.6 mm (1.125×1.125")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1254,57 $Supplier Number: 807656026527
- Warning labels - Stop SignPlasticWhite on red3"38.1×38.1 mm (1.5″ × 1.5")1000 labels/roll (1 label across)Log in pour la disponibilitéRoll of 1328,22 $Supplier Number: 807656026619
- Warning labels - Non HazardousPlasticBlack on mint3"44.5×19.1 mm (1.75×0.75")250 labels/roll (1 label across)Log in pour la disponibilitéRoll of 161,27 $Supplier Number: 807656026503
- Warning labels - Toxic Warning SymbolPlasticBlack on red3"44.5×19.1 mm (1.75×0.75")250 labels/roll (1 label across)Log in pour la disponibilitéRoll of 161,17 $Supplier Number: 807656026497
- Warning labels - BiohazardPlasticBlack on red3"50.8×50.8 mm (2×2")100 labels/roll (1 label across)Log in pour la disponibilitéRoll of 185,05 $Supplier Number: 807656083940
- Discontinued-Supplier Number: -