Sharpie® Sterile Markers, Veltek Associates
Catalog # 76308-430
Supplier: Veltek Associates
Specifications
- Sterile:Sterile
- Color:Black
- Description:Sharpie® sterile marker
- Cat. no.:76308-430
Specifications
About this item
These gamma irradiated sterile sharpie markers compatible with line of cleanroom documentation forms, logbooks, tags, and labels and for CleanPrint 10 synthetic writing substrate.
- Fine tip
- Permanent black ink
Packaging: Individually double bagged.