Klinger Beryl Crystal Model
Beryl Crystal Model, 36×24×12", 12 lbs.
Catalog # 470311-030
Supplier: Klinger
Fournisseur #: KS8081
Jump to:
Product Details & Documents
This model is hand made in the USA by Klinger Educational Products.
- Contains 256 - 1" balls
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Spécifications
- Désignation:Beryl Crystal Model
- Dimensions:36×24×12"
- Weight:12 lbs.