Klinger Wurtzite Crystal Model
Wurtzite Crystal Model, 24×24×12", 6 lbs.
Catalog # 470311-020
Supplier: Klinger
Fournisseur #: KS8074
Jump to:
Product Details & Documents
This model is hand made in the USA by Klinger Educational Products.
- Contains 39 - 1" balls
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Spécifications
- Désignation:Wurtzite Crystal Model
- Dimensions:24×24×12"
- Weight:6 lbs.