Perforated Spoons | Avantor
Perforated Spoons
Perforated spoon, bowl Ø: 2 cm (0.8"), 15 cm (5.9"), Stainless steel
Catalog # 77663-018
Fournisseur #:  501997
Perforated Spoons
Catalog # 77663-018
Perforated Spoons
Perforated spoon, bowl Ø: 2 cm (0.8"), 15 cm (5.9"), Stainless steel
Price
$73.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perforated spoon made of stainless steel.
Spécifications
  • Désignation:
    Perforated spoon, bowl Ø: 2 cm (0.8")
  • Longueur:
    15 cm (5.9")
  • Matière:
    Stainless steel