U-Shaped Spargers, Analytical Purge and Trap, Kimble Chase
These U-shaped fritted spargers are designed for use with O-I analytical purge and trap sample concentrators.
- Recommended for potable water and wastewater samples
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together