Q-SWAB™ Dry Swab with Transport Tube, Simport
Sterile specimen collection swabs.
- Polypropylene tubes are translucent
- Cap is equipped with a silicone washer
- Non-graduated
The Q-Swab™ design is self-standing, but with round bottom. The polyester swab holds securely inside the screw cap, making it ideal for specimen collection and protection from contamination. The tube material make it easy to see all contents. The cap features with washer fitted inside to ensure a positive seal at any temperature.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together