The EcoSense® ODO200M is an optical-based dissolved oxygen meter ideal for DO sampling. Sensors are EPA approved for wastewater compliance reporting of DO.
- Waterproof micro-USB port for transferring data to PC; no software required
- Holds 250 data sets; no need to write down data
- Real-time clock for date/time stamp of saved data
- User-selectable recalibration prompt provides reminder to calibrate
- Automatic temperature compensation; manual input for salinity and pressure compensation
- IP 67 waterproof housing
- Replaceable optical sensor cap with 12 to 18 month life
- Auto shutoff function after 30 minutes of inactivity
- User calibratable and replaceable
- Sampling
Compared to the EcoSense® ODO200 , the ODO200M features extended memory and simple data transfer with a micro-USB port. Optical benefits include no stirring requirement, no warm-up time, no electrolyte solutions, and no electrode maintenance.
The ODO200M is designed for quick, accurate results in an economical platform. The convenient optical dissolved oxygen sensor and the simple, compact handheld design allow the ODO200M to be used in many applications.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76300-000
- 76300-002
- 76300-004
- 76300-006
- DO range:
- 0 - 20 mg/l
- 0 - 20 mg/l
- 0 - 20 mg/l
- 0 - 20 mg/l
- DO saturation:
- 0 - 200 %
- 0 - 200 %
- 0 - 200 %
- 0 - 200 %
- DO probe type:
- Optical
- Optical
- Optical
- Optical
- Temperature range:
- 0...50 °C
- 0...50 °C
- 0...50 °C
- 0...50 °C
- Power supply:
- One 9 V battery (included)
- One 9 V battery (included)
- One 9 V battery (included)
- One 9 V battery (included)
- Memory:
- 250 data set reviewable memory
- 250 data set reviewable memory
- 250 data set reviewable memory
- 250 data set reviewable memory
- Operating temperature range:
- 0...50 °C (32...122 °F)
- 0...50 °C (32...122 °F)
- 0...50 °C (32...122 °F)
- 0...50 °C (32...122 °F)