EcoSense® EC300M-Conductivity, TDS, Salinity, Temperature Instrument with Extended Memory, YSI
Fournisseur: YSI
Certificats
À propos de cet article
The EC300M offers convenient measurement of conductivity, specific conductance, salinity, total dissolved solids, and temperature in many field applications. Compared to the EcoSense® EC300A the EC300M features extended memory and simple data transfer with a micro-USB port.
- Holds 250 data sets; no need to write down data
- Waterproof micro-USB port for transferring data to PC; no software required
- Real-time clock for date/time stamp of saved data
- User-selectable recalibration prompt provides reminder to calibrate
- Automatic temperature compensation
- Super-stable, four-electrode conductivity cell designed for the field
- IP 67 waterproof housing
- Auto shutoff function after 30 minutes of inactivity
- User calibratable and replaceable
The EC300M is designed for quick, accurate results in an economical platform. With a one-year instrument and electrode warranty, waterproof case and ease of use, the EC300M will fit your needs.
Units: Conductivity: ms/cm or µs/cm; Salinity: ppt; Total Dissolved Solids (TDS): mg/l; Temperature: °C
Ordering information: The following EC300M purchase options are available: EC300M instrument only; or EC300M instrument, hard-sided transport case, and probe with 1 meter Field Cable; or EC300M instrument, hard-sided transport case, and probe with 4 meter field cable; or EC300M instrument, hard-sided transport case, and probe with 10 meter field cable. One year instrument and probe/cable assembly warranty.
Spécifications
- Catalog No:
- 76299-982
- 76299-984
- 76299-986
- 76299-988
- Model:
- EC300M
- EC300M
- EC300M
- EC300M
- Parameters:
- Conductivity, Salinity, TDS, Temperature
- Conductivity, Salinity, TDS, Temperature
- Conductivity, Salinity, TDS, Temperature
- Conductivity, Salinity, TDS, Temperature
- Conductivity range:
- 0 - 200.0 mS/cm
- 0 - 200.0 mS/cm
- 0 - 200.0 mS/cm
- 0 - 200.0 mS/cm
- Salinity range:
- 0.0 - 70.0 ppt
- 0.0 - 70.0 ppt
- 0.0 - 70.0 ppt
- 0.0 - 70.0 ppt
- Temperature range:
- –10...90 °C (14...194 °F)
- –10...90 °C (14...194 °F)
- –10...90 °C (14...194 °F)
- –10...90 °C (14...194 °F)
- Power supply:
- One 9 V battery (included)
- One 9 V battery (included)
- One 9 V battery (included)
- One 9 V battery (included)
- Operating temperature range:
- 0...50 °C (32...122 °F)
- 0...50 °C (32...122 °F)
- 0...50 °C (32...122 °F)
- 0...50 °C (32...122 °F)
- Memory:
- 250 data set reviewable memory
- 250 data set reviewable memory
- 250 data set reviewable memory
- 250 data set reviewable memory