Measuring Spoon, Hach
Supplier:   Hach
Measuring Spoon, Hach
Supplier:  Hach
Certificates
Measuring Spoon, Hach
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Measuring spoon features a molded plastic cup. Weight determination is based on sodium chloride.
Product Family Options
Informations sur le produit
  • Matière
    Capacité
    Availability
    Price
  • Plastic
    0.1 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.43
    $11.43
    Each
       $11.43
    Supplier Number: 51100
  • Plastic
    1.0 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14.28
    $14.28
    Each
       $14.28
    Supplier Number: 51000