G-Tek® KEV™ Seamless Knit Kevlar® Gloves with Crinkle Latex Grip, Protective Industrial Products
Supplier: PIP
Certificates
About this item
Ideal for material handling, glass handling operations, fishing industries, sanitation, general maintenance, woodworking, waste handling and recycling.
- Superior cut resistance
- Knit wrist helps prevent dirt and debris from entering the glove
- Color coded hems for easy size identification
- Launderable for extended life and to reduce replacement costs
Seamless knit construction protects hands without sacrificing comfort or dexterity. The Kevlar® fiber shell provides high tensile strength relative to its weight, is inherently cut resistant and will not melt, ignite or conduct electricity.
The latex crinkle coating on the palms and fingers is designed to channel away liquids to allow a better handling in heavy duty wet and dirty conditions. The thicker coating provides additional protection for cut and puncture applications.
Certifications: EN 388: 3544. ASTM F2992-15 ANSI Cut Level A4. ASTM F1790-05 ANSI Cut Level 4.
Specifications
- Catalog No:
- 75814-468
- 75814-470
- 75814-462
- 75814-466
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Material:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Finish:
- Crinkle Latex Palm and Finger Coating
- Crinkle Latex Palm and Finger Coating
- Crinkle Latex Palm and Finger Coating
- Crinkle Latex Palm and Finger Coating
- Cuff style:
- Elastic
- Elastic
- Elastic
- Elastic