CleanTeam® Medium Weight Cotton Lisle Inspection Gloves, Protective Industrial Products
: PIP
Designed to minimize contamination of products from human hands. Ideal for inspection and electronics assembly.
- Constructed in a two piece, reversible pattern using 100% cotton fabric
- Can be used as liners under disposable and unsupported liquid proof gloves
Unhemmed cuff style.
: Made from component materials which comply with Federal regulations for food contact. 21CFR, 170-99.
- Catalog No:
- 20000-533
- 20000-537
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Material:
- Cotton
- Cotton
Frequently Bought Together