Optimum Growth® Sampling and Multiport Flasks
Optimum Growth® Sampling and Multiport Flasks
Optimum Growth® Sampling and Multiport Flasks
Certificates
Optimum Growth® Sampling and Multiport Flasks
Optimum Growth® Sampling and Multiport Flasks
9 Options Available Below
Product Details & Documents
Thomson’s Optimum Growth® Sampling and multiport flasks are engineered for contamination-free sampling, aseptic feeding, and sterile media transfer, making them ideal for seed train development, upstream bioprocessing, and closed-system cell culture maintenance. These flasks eliminate the need for rocker bioreactors, transfer containers, and tube fusing setups while maintaining sterility and shaker compatibility throughout the workflow.
  • Available in multiple sizes
  • One-way sampling valve allows safe and aseptic sampling while the flask remains in the shaker or on the benchtop
  • Streamlines workflows and minimizes time, contamination risk, and handling complexity
  • Closed system designs helps maintain sterility throughout use, reducing contamination risk and supporting GMP compliance
Together, these flasks deliver a powerful combination of ease-of-use, contamination control, and operational flexibility, making them a leading choice for modern upstream bioprocessing workflows.
Thomson’s Optimum Growth® Sampling Flasks feature integrated one-way valves that enable aseptic sampling directly in the shaker or on the benchtop, helping reduce contamination risk and improve efficiency, especially in high-throughput environments. These flasks are available in multiple sizes with integrated one-way sampling valves, allowing aseptic sampling without removing the cap. Their design streamlines workflows and minimizes time, contamination risk, and handling complexity. The 0.2 µm PTFE vented caps support sterile gas exchange and maintain culture integrity.
Thomson’s Optimum Growth® Multiport flasks offer integrated ports that enable closed-system feeding, sampling, and media transfer, supporting sterile workflows without the need to open the flask, even during extended culture periods. These flasks are available in multiple sizes with a one-way sampling valve that allows safe and aseptic sampling while the flask remains in the shaker or on the benchtop. A two-way Luer lock port supports sterile feeding, inoculation, and direct media transfer to larger vessels with no tube fusing required, as media can be added or withdrawn through integrated ports. Their design eliminates the need for cell bags up to 50 L, reducing cost and complexity. Compatible with multi-day batch feeding in shaker environments, multiport flasks include sterile, pre-attached tubing for plug-and-play integration into cleanroom workflows.
Documents
Product Family Options
Multiport flasks
Product Information
  • Désignation
    Capacité
    Color
    Sterility
    Cap Type
    Longueur Tubes
    Availability
    Price
  • Multiport Optimum Growth™ flask with threaded top, dual port vent cap, Male Luer lock connections, 381 µl sample tubing, C-Flex® 16 tubing (¹/₄"Ext.Ø.×¹/₈"Int.Ø)
    250 ml
    Translucent
    Sterile
    Screw cap
    24" (609.6 mm)
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $2,020.00
    $2,020.00
    Case of 40
       $2,020.00
    Minimum order quantity is 1
    Supplier Number: 931111-DP
  • Multiport Optimum Growth™ flask with threaded top, dual port vent cap, Male Luer lock connections, 381 µl sample tubing, C-Flex® 16 tubing (¹/₄"Ext.Ø.×¹/₈"Int.Ø)
    500 ml
    Translucent
    Sterile
    Screw cap
    24" (609.6 mm)
    Log in for availability
    Log in for availability
    Case of 15
    Case of 15 - $1,030.00
    $1,030.00
    Case of 15
       $1,030.00
    Minimum order quantity is 1
    Supplier Number: 931112-DP
  • Multiport Optimum Growth™ flask with threaded top, dual port vent cap, Male Luer lock connections, 381 µl sample tubing, C-Flex® 16 tubing (¹/₄"Ext.Ø.×¹/₈"Int.Ø)
    125 ml
    Translucent
    Sterile
    Screw cap
    24" (609.6 mm)
    Log in for availability
    Log in for availability
    Case of 30
    Case of 30 - $1,620.00
    $1,620.00
    Case of 30
       $1,620.00
    Minimum order quantity is 1
    Supplier Number: 931110-DP
  • Multiport Optimum Growth™ flask with threaded top, vent cap, Male Luer lock connections, 381 µl sample tubing, C-Flex® 16 tubing (¹/₄"Ext.Ø.×¹/₈"Int.Ø)
    1.6 L
    Translucent
    Sterile
    Screw cap
    24" (609.6 mm)
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $3,600.00
    $3,600.00
    Case of 12
       $3,600.00
    Minimum order quantity is 1
    Supplier Number: 931113-PORT-TRT
  • Multiport Optimum Growth™ flask with threaded top, vent cap, Male Luer lock connections, 381 µl sample tubing, C-Flex® 16 tubing (¹/₄"Ext.Ø.×¹/₈"Int.Ø)
    5 L
    Translucent
    Sterile
    Screw cap
    24" (609.6 mm)
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,780.00
    $1,780.00
    Case of 4
       $1,780.00
    Minimum order quantity is 1
    Supplier Number: 931116-PORT-TRT-F
Sampling flasks
Product Information
  • Désignation
    Capacité
    Color
    Sterility
    Cap Type
    Longueur Tubes
    Availability
    Price
  • Optimum Growth™ flask with sampling port, threaded top, sampling vent cap, 0.2 µm PTFE ventilation, Male Luer lock connection, 163 µl sample tubing
    125 ml
    Translucent
    Sterile
    Screw cap
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $2,120.00
    $2,120.00
    Case of 50
       $2,120.00
    Minimum order quantity is 1
    Supplier Number: 931110-SP
  • Optimum Growth™ flask with sampling port, threaded top, sampling vent cap, 0.2 µm PTFE ventilation, Male Luer lock connection, 128 µl sample tubing
    250 ml
    Translucent
    Sterile
    Screw cap
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $2,230.00
    $2,230.00
    Case of 50
       $2,230.00
    Minimum order quantity is 1
    Supplier Number: 931111-SP
  • Optimum Growth™ flask with sampling port, threaded top, sampling vent cap, 0.2 µm PTFE ventilation, Male Luer lock connection, 313 µl sample tubing
    500 ml
    Translucent
    Sterile
    Screw cap
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $1,570.00
    $1,570.00
    Case of 25
       $1,570.00
    Minimum order quantity is 1
    Supplier Number: 931112-SP
  • Optimum Growth™ flask with sampling port, threaded top, sampling vent cap, 0.2 µm PTFE ventilation, Male Luer lock connection, 381 µl sample tubing
    5 L
    Translucent
    Sterile
    Screw cap
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $953.00
    $953.00
    Case of 4
       $953.00
    Minimum order quantity is 1
    Supplier Number: 931116-PORT-E
Accessories
Product Information
  • Catalog #
    Sterility
    Désignation
    Availability
    Price
  • Sterile
    Optimum Growth® vent caps for 125 ml flasks
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $641.00
    $641.00
    Case of 50
       $641.00
Product Information
  • Catalog #
    Sterility
    Désignation
    Availability
    Price
  • Sterile
    Optimum Growth® vent caps for 250 ml flasks
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $641.00
    $641.00
    Case of 50
       $641.00
Product Information
  • Catalog #
    Sterility
    Désignation
    Availability
    Price
  • Sterile
    Optimum Growth® vent caps for 500 ml flasks
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $387.00
    $387.00
    Case of 25
       $387.00
Product Information
  • Catalog #
    Sterility
    Désignation
    Availability
    Price
  • Sterile
    Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $603.00
    $603.00
    Case of 24
       $603.00
Product Information
  • Catalog #
    Sterility
    Désignation
    Availability
    Price
  • Sterile
    Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $415.00
    $415.00
    Case of 24
       $415.00
Product Information
  • Catalog #
    Sterility
    Désignation
    Availability
    Price
  • Sterile
    Optimum Growth® 1.6 L flask and bioreactor exhaust caps
    Log in for availability
    Log in for availability
    Case of 8
    Case of 8 - $2,210.00
    $2,210.00
    Case of 8
       $2,210.00
amed Group
Product Information
  • Catalog #
    Matière
    Sterility
    Désignation
    Availability
    Price
  • Polypropylene
    Sterile
    Rapid Clear® cap 3000
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,700.00
    $1,700.00
    Case of 4
       $1,700.00
Accessories
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • 8-position carrier for 125 ml Optimum Growth® flasks
    8-position carrier for 125 ml Optimum Growth® flasks
    76808-360
    10.75×5"
    Log in for availability
    Each - $1,210.00
Product Information
  • Catalog #
    Dimensions
    Availability
    Price
  • 8-position carrier for 250 ml Optimum Growth® flasks
    8-position carrier for 250 ml Optimum Growth® flasks
    76808-362
    13.4×6"
    Log in for availability
    Each - $1,210.00
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Stand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasks
    Stand with ring for inverting 1.6 and 2.8 L Optimum Growth® flasks
    76808-514
    5" ring
    Log in for availability
    Each - $340.00
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Stand with ring for inverting 5 L Optimum Growth® flasks
    Stand with ring for inverting 5 L Optimum Growth® flasks
    76808-510
    7" ring
    Log in for availability
    Each - $340.00
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Ring only for inverting 1.6 and 2.8 L Optimum Growth® flasks
    Ring only for inverting 1.6 and 2.8 L Optimum Growth® flasks
    76808-516
    5" ring
    Log in for availability
    Each - $170.00
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Ring only for inverting 5 L Optimum Growth® flasks
    Ring only for inverting 5 L Optimum Growth® flasks
    76808-512
    7" ring
    Log in for availability
    Each - $170.00
Product Information
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 500 ml flasks
    Optimum Growth® vent caps for 500 ml flasks
    76808-548
    Sterile
    Log in for availability
    Case of 25 - $387.00
Product Information
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® 1.6 L flask and bioreactor exhaust caps
    Optimum Growth® 1.6 L flask and bioreactor exhaust caps
    76808-330
    Sterile
    Log in for availability
    Case of 8 - $2,210.00
Product Information
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks
    Optimum Growth® vent caps for 1.6 L, 2.8 L, and 5 L flasks
    76808-554
    Sterile
    Log in for availability
    Case of 24 - $603.00
Product Information
  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Rapid Clear® cap 3000
    Rapid Clear® cap 3000
    76808-350
    Polypropylene
    Sterile
    Log in for availability
    Case of 4 - $1,700.00
Product Information
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 125 ml flasks
    Optimum Growth® vent caps for 125 ml flasks
    76808-544
    Sterile
    Log in for availability
    Case of 50 - $641.00
Product Information
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® vent caps for 250 ml flasks
    Optimum Growth® vent caps for 250 ml flasks
    76808-546
    Sterile
    Log in for availability
    Case of 50 - $641.00
Product Information
  • Catalog #
    Sterility
    Availability
    Price
  • Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks
    Optimum Growth® low evaporation caps for 1.6, 2.8, and 5 L flasks
    76808-354
    Sterile
    Log in for availability
    Case of 24 - $415.00