Klinger Graphite ll (Hexagonal) Crystal Model | Avantor
Klinger Graphite ll (Hexagonal) Crystal Model
Graphite ll Crystal Model, 24×20×14", 4 lbs.
Catalog # 470311-132
Supplier:   Klinger
Fournisseur #:  KS8035
Klinger Graphite ll (Hexagonal) Crystal Model
Catalog # 470311-132
Supplier:  Klinger
Klinger Graphite ll (Hexagonal) Crystal Model
Klinger Graphite ll (Hexagonal) Crystal Model
Graphite ll Crystal Model, 24×20×14", 4 lbs.
Price
$624.55
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
This model is hand made in the USA by Klinger educational products.
  • Permanent structures
  • Made with grade A materials
The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Spécifications
  • Désignation:
    Graphite ll Crystal Model
  • Dimensions:
    24×20×14"
  • Weight:
    4 lbs.