Klinger Copper Crystal Model
Copper Crystal Model, 12×12×12", 4 lbs.
470311-126
: Klinger
: KS8025
This model is hand made in the USA by Klinger educational products.
- Permanent structures
- Made with grade A materials
The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
- Désignation:Copper Crystal Model
- Dimensions:12×12×12"
- Weight:4 lbs.