Screw Caps for Internal Thread Sample Tubes, Simport Scientific
Supplier: Simport Scientific
Certificates
About this item
These PP screw caps are to be used with T500 series internal thread sample tubes.
- Silicone washer seal
- Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
Ordering information: Tubes T500 series sold separately.
Frequently Bought Together