Micro-Touch® Micro-Thin™ Nitrile Examination Gloves
Nitrile, powder-free, medical exam glove with a thin film formulation for excellent tactile sensitivity.
Thin nitrile formulation offers excellent tactile sensitivity
- Textured surface for sure handling of materials and instruments
- Smart packaging
- 300 gloves per box reduces restocking
Offering heightened sensitivity, this non-sterile glove is made with a thinner formulation to provide excellent tactile sensitivity and dexterity, suitable for a variety of clinical procedures, including palpating veins and starting IVs. The nitrile formulation also provides exceptional comfort, fit, and feel. The textured surface of the glove promotes sure handling of materials and instruments. Designed to provide specialized protection, this glove has been tested against chemotherapy drugs and is food handling compliant.
: ASTM D6319, CE 0493, medical device regulation (EU) 2017/745 class I, FDA21 CFR 177-2600-US food contact approved, food type: rubber, ISO 9001 and 13485.
: Gloves are packaged 300 pieces per dispenser, ten dispensers per box.
Nitrile, powder-free, medical exam glove with a thin film formulation for excellent tactile sensitivity.
Thin nitrile formulation offers excellent tactile sensitivity
- Textured surface for sure handling of materials and instruments
- Smart packaging
- 300 gloves per box reduces restocking
Offering heightened sensitivity, this non-sterile glove is made with a thinner formulation to provide excellent tactile sensitivity and dexterity, suitable for a variety of clinical procedures, including palpating veins and starting IVs. The nitrile formulation also provides exceptional comfort, fit, and feel. The textured surface of the glove promotes sure handling of materials and instruments. Designed to provide specialized protection, this glove has been tested against chemotherapy drugs and is food handling compliant.
: ASTM D6319, CE 0493, medical device regulation (EU) 2017/745 class I, FDA21 CFR 177-2600-US food contact approved, food type: rubber, ISO 9001 and 13485.
: Gloves are packaged 300 pieces per dispenser, ten dispensers per box.
- Catalog No:
- CA75839-358
- 77661-322
- CA75839-360
- 75839-358
- CA75839-362
- 75839-360
- CA75839-364
- 75839-362
- 75839-364
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff length:
- Standard
- Standard
- Standard
- Standard
- Standard
- Standard
- Standard
- Standard
- Standard
- Finger thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Palm thickness:
- 2.4 mil
- 2.4 mil
- 2.4 mil
- 2.4 mil
- 2.4 mil
- 2.4 mil
- 2.4 mil
- 2.4 mil
- 2.4 mil
- FDA regulated:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Food contact approved:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- ASTM No.:
- D6319
- D6319
- D6319
- D6319
- D6319
- D6319
- D6319
- D6319
- D6319
Product Family Options
- Taille du gantGlove ColorLongueur
- : 6034311
Specifications
Cat. No.CA75839-358Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - XSBlue230 mm (9.1")Box of 300$35.20Case of 3000$351.97: 6034310
Specifications
Cat. No.77661-322Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - : 6034312
Specifications
Cat. No.CA75839-360Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - SBlue230 mm (9.1")Box of 300$35.20Case of 3000$351.97: 6034311
Specifications
Cat. No.75839-358Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - : 6034313
Specifications
Cat. No.CA75839-362Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - MBlue230 mm (9.1")Box of 300$35.20Case of 3000$351.97: 6034312
Specifications
Cat. No.75839-360Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - : 6034314
Specifications
Cat. No.CA75839-364Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - LBlue230 mm (9.1")Box of 300$35.20Case of 3000$351.97: 6034313
Specifications
Cat. No.75839-362Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319 - XLBlue230 mm (9.1")Box of 300$35.26Case of 3000$352.64: 6034314
Specifications
Cat. No.75839-364Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatex-freePowdered/Powder-freePowder-freeFinishTextured fingersCuff styleBeadedCuff lengthStandardFinger thickness3.9 milPalm thickness2.4 milFDA regulated+SterilityNon sterileFood contact approved+ASTM No.D6319