À propos de cet article
A sensitive detection method for measuring the free acid form of fluprostenol.
- Measure the free acid form of fluprostenol
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure fluprostenol levels down to 16 pg/ml
- Colorimetric reading at 405 to 420 nm
Fluprostenol is a metabolically stable analog of PGF2α with potent FP receptor agonist activity. The isopropyl ester of fluprostenol has recently been approved for use as an ocular hypotensive drug for the treatment of glaucoma and is sold under the Alcon trade name Travoprost. Acting as a prodrug, Fluprostenol isopropyl ester is converted by esterase activity in the cornea to yield the corresponding active free acid, (+)-fluprostenol. Fluprostenol is also used in the veterinary field for the induction of estrus in livestock. Cayman’s Fluprostenol ELISA is a sensitive detection method for measuring the free acid form of fluprostenol. The assay also detects the isopropyl ester form of fluprostenol, but with less sensitivity. Samples containing a mixture of the isopropyl ester and free acid should be purified and the compounds separated prior to analysis. The assay has been validated for use in aqueous humor samples.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Mid point:110 pg/ml
- Detection range:7.8 - 1000 pg/ml
- Sensitivity:16 pg/ml
- Incubation time:18 hr