ContactUsLinkComponent
PPARδ Transcription Factor Assay Kit, Cayman Chemical Company
Catalog # 75816-694
CAS Number:  
undefined
PPARδ Transcription Factor Assay Kit, Cayman Chemical Company
Catalog # 75816-694
Supplier Number:  10006914-96
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    PPARδ Transcription Factor Assay Kit, 96 Strip Wells
  • Cat. no.:
    75816-694
  • Supplier No.:
    10006914-96
Specifications
About this item
A 96-well assay for measurement of PPARδ DNA binding activity.
  • A sensitive, non-radioactive method of detecting PPARδ from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
  • Plate-based colorimetric measurement (405 to 420 nm)
  • Nuclear extraction kit is available to aid in the isolation of nuclear and cytoplasmic fractions from cell lysates or tissue homogenates
PPARs are ligand-activated transcription factors belonging to the large superfamily of nuclear receptors. PPARα primarily activates genes encoding proteins involved in fatty acid oxidation, while PPARγ primarily activates genes directly involved in lipogenic pathway and insulin signaling. Members of the PPAR family are important direct targets of many antidiabetic and hypolipidemic drugs. Cayman’s PPAR Transcription Factor Assays are a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
Delivery: Kit contains assay buffer, reagent A, positive control, binding buffer, primary antibody, wash buffer concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover sheet, developing and stop solution.