High Viscosity Standards, Cannon Instruments | Avantor
High Viscosity Standards, Cannon Instruments
Supplier:   Cannon
High Viscosity Standards, Cannon Instruments
Supplier:  Cannon
Certificates
High Viscosity Standards, Cannon Instruments
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These certified standards are useful for calibration and verification procedures.
  • For asphalt and polymer applications
Certifications: Meets the specifications of ASTM D2170 and ASTM D2171.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Capacité de flacon
    Availability
    Price
  • High viscosity standard, N30000
    3.8 L
    (20°C) 120000 mPa·s
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1US Gal)
    Each (1US Gal) - $1,187.84
    $1,187.84
    Each (1US Gal)
       $1,187.84
    Supplier Number: 9727-E25.128
  • High viscosity standard, N30000
    5 Gal.
    (20°C) 120000 mPa·s
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20l)
    Each (20l) - $4,454.91
    $4,454.91
    Each (20l)
       $4,454.91
    Supplier Number: 9727-E25.500
  • High viscosity standard, N30000
    500 ml
    (20°C) 120000 mPa·s
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500ml)
    Each (500ml) - $192.17
    $192.17
    Each (500ml)
       $192.17
    Supplier Number: 9727-E25.016
  • High viscosity standard, N30000
    950 ml
    (20°C) 120000 mPa·s
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1l)
    Each (1l) - $329.36
    $329.36
    Each (1l)
       $329.36
    Supplier Number: 9727-E25.032
  • High viscosity standard, N2700000SP
    55 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (55ml)
    Each (55ml) - $416.07
    $416.07
    Each (55ml)
       $416.07
    Supplier Number: 9727-E42