Rectangular Semi-Demountable Cell Windows, PerkinElmer | Avantor
Rectangular Semi-Demountable Cell Windows, PerkinElmer
Supplier:   PerkinElmer
Rectangular Semi-Demountable Cell Windows, PerkinElmer
Supplier:  PerkinElmer
Certificates
Rectangular Semi-Demountable Cell Windows, PerkinElmer
Rectangular Semi-Demountable Cell Windows, PerkinElmer
3 Options Available Below
Product Details & Documents
Rectangular windows make the demountable cell becomes a semi-demountable cell. It can be used either as a demountable cell or as a sealed cell for high-viscosity liquids or mull samples.
  • Available in a variety of window materials
  • Windows come in pairs
Combining two different requirements into one cell allows a myriad of different sampling applications.
Ordering information: Semi-demountable Cell can be ordered with the universal demountable cell, windows and spacers.
Product Family Options
Informations sur le produit
  • Désignation
    Hauteur
    Épaisseur
    Largeur
    Availability
    Price
  • Sodium Chloride Cell Windows
    41 mm
    4 mm
    23 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $652.62
    $652.62
    Pack of 2
       $652.62
    Supplier Number: L1271191
  • Potassium Bromide Cell Windows
    41 mm
    4 mm
    23 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $614.58
    $614.58
    Pack of 2
       $614.58
    Supplier Number: L1271192
  • Calcium Fluoride Cell Windows
    41 mm
    4 mm
    23 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $1,323.23
    $1,323.23
    Pack of 2
       $1,323.23
    Supplier Number: L1271193