ContactUsLinkComponent
PELTOR™ X3 Over-the-Head Earmuffs, 3M™
  76308-688
 :  3M
undefined
PELTOR™ X3 Over-the-Head Earmuffs, 3M™
  76308-688
 :  3M
 :  7000104072
  • NRR:
    28 dB
  • Color:
    Black/Red
  • Description:
    Over-the-head earmuffs
  • Cat. no.:
    76308-688
This lightweight earmuff is designed for protection against high level industrial noise and other loud sounds.
  • X3A features a newly designed spacer to help improve attenuation without the need for a much larger cup or double cup design
  • Electrically insulated wire headband; dielectric
Recommended for painting, cleaning, assembly and mechanical work.