Specifications
- For Use With:Flat mops, rectangular slim T buckets
- Description:Down-press flat head mop wringer
- Dimensions:51 L×11.4W×25 D cm (20.5×4.5×10")
- Material:Stainless steel 304, electropolished, polyketone
- Cat. no.:76299-864
Specifications
About this item
This lightweight down press wringer is ideal for consistent flat head mop wringing with minimal effort.
- Unique design allows for ergonomic, no operator bend, one hand action
- Mop handle slides into the wringer grip and provides the leverage to create wringing pressure
- Minimizes dripping and over saturation of flat head mops, removing much more liquid than a sieve wringer
- Lightweight and durable, the wringer is a combination of molded Polyketone and electropolished stainless steel
- Simple storage and cleaning; wringer easily removes from buckets
- No cart is necessary to hold the wringer
- Autoclavable and compatible with strong disinfectants and cleaning solutions
For applications where traditional wetting and wringing methods are considered more effective, Micronova has introduced the MegaWringer™. This modular, downward press wringer will efficiently wring flat heads with minimum effort. Operators use the leverage of the mop handle to create the wringing pressure, doing away with the need to constantly bend and pull while cleaning. This guards against operator wrist and back strain.
A combination of electropolished stainless steel and polyketone, the MegaWringer™ fits most rectangular bucket systems. Plastic and stainless steel, the unit weighs less than other downward press wringing mechanisms and is compact and easily transported and stored.
MegaWringer™ easily clamps directly to the Micronova B-7 series bucket in a single, double, or triple bucket configuration. Works with the BucketBinder Multi-Bucket System or SlimT™ Double Bucket System.