pH Buffers, YSI
À propos de cet article
pH buffers for pH probe calibration.
Each buffer case contains 6 individual pints (plastic). Buffer cases are available with a single buffer type or with an assortment of buffers (2 pints of pH 4, 7, and 10 buffers).
3 Options Available Below
Product Details & Documents
pH buffers for pH probe calibration.
Each buffer case contains 6 individual pints (plastic). Buffer cases are available with a single buffer type or with an assortment of buffers (2 pints of pH 4, 7, and 10 buffers).
Documents
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- DésignationpH AccuracyPackedTaillepHColorAvailabilityPrice
- pH Buffer±0.01Box of 6 Pints475 ml4redEach$42.52Pack of 6$255.12Supplier Number: 3821
- pH Buffer±0.01Box of 6 Pints475 ml7yellowEach(1/Itm/Bx)$43.96Pack of 6 (1/ Itm/Bx)$263.76Supplier Number: 003822
- pH Buffer±0.01Box of 6 Pints475 ml10BlueEach$42.52Pack of 6$255.12Supplier Number: 03823