Wizard™ CompactDry™ Reader, Hardy Diagnostics
Wizard™ CompactDry™ Reader
Catalog # CA76177-196
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: CDR1
Jump to:
Product Details & Documents
Bring automation, efficiency and data security into your lab.
- Automatically reads CompactDry™ plates in less than four seconds – stand alone system, no PC required
- Counts and differentiates colonies based on the CompactDry™ type
- Indexes cassette directly under the imaging camera for rapid reading
- Accurately reads up to 150 colonies per plate
- Accepts individual plates or groups of two, three, or four plates so you can keep your dilution series together - no need to seperate plates
- Unit is suited for ambidextrous operation; plates can be scanned from the left or right side
- Digitalized plate image can be saved to USB or exported to the user's LIMS program or included software management program
The Wizard™ CompactDry™ Reader is a fast, easy, and accurate way to read and record your CompactDry™ results. The Wizard™ has selectable modes in the configuration menu designed to read the different types of CompactDry™ plates.
Informations sur les accessoires: Available accessories include USB wireless keyboard, USB barcode scanner, and USB thermal printer. Optional keyboard accessory for Wizard™ CompactDry™ Reader connects to Wizard™ by wireless USB dongle. Barcode reader for The Wizard™ CompactDry™ Reader connects via USB. Thermo Printer Option for the Wizard™ CompactDry™ Reader connects via USB.
Spécifications
- Désignation:Wizard™ CompactDry™ Reader
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Thermal printer option for The Wizard™ CompactDry™ Reader, connects via USBThermal printer option for The Wizard™ CompactDry™ Reader, connects via USBCA76177-152Log in pour la disponibilitéEach - $2,170.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Keyboard for Wizard™ CompactDry™ Reader, connects to Wizard™ by wireless USB dongleKeyboard for Wizard™ CompactDry™ Reader, connects to Wizard™ by wireless USB dongleCA76177-156Log in pour la disponibilitéEach - $295.00