Micro Funnel, Hach® | Avantor
Micro Funnel, Hach®
Micro Funnel, 35 mm
Catalog # CA25843-35
Supplier:   Hach
Fournisseur #:  2584335
Micro Funnel, Hach®
Catalog # CA25843-35
Supplier:  Hach
Micro Funnel, Hach®
Micro Funnel, 35 mm
Price
$14.12
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for micro work, filling burets, manometers, and small bottles.
  • Polypropylene construction
  • Suitable for micro work
  • Steam autoclavable at 121 °C (250 °F)
Spécifications
  • Désignation:
    Micro Funnel
  • Top I.D.:
    35 mm