VWR® Urethane Lab Stools | Avantor
VWR® Urethane Lab Stools
Supplier:   VWR International
VWR® Urethane Lab Stools
Supplier:  VWR International
Certificates
VWR® Urethane Lab Stools
4 Options Available Below
Product Details & Documents
Urethane lab stools have easy-to-clean seats which resist many chemicals, water, paint and more, combined with ergonomic functionality that make them a popular choice of laboratory professionals.
  • Other components include an easy-to-use ergonomic task control with seat height and back tilt adjustments and 26" Ø black composite base
  • Designed for laboratory applications, including those in healthcare, education and industrial settings
  • Backed by a 10-year warranty (lifetime on pneumatic)
Lab chair line features a refined yet pragmatic collection of chairs and stool designed for today laboratory professionals, applications and settings. The combination of european design with proven ergonomic functionality and engineering results in a focused set of models that meet real-world lab seating challenges.
Certifications: Meets ANSI/BIFMA standards for durability and safety.
Caution: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76308-804
  • 76308-806
  • 76308-808
  • 76308-810
  • Model:
  • 76308-804
  • 76308-806
  • 76308-808
  • 76308-810
  • For use with:
  • Medium height benches (32.75 - 42")
  • Medium height benches (32.75 - 42")
  • Low benches (27-32")
  • Low benches (27-32")
  • Seat/back color:
  • Black
  • Black
  • Black
  • Black
  • Seat/back material:
  • Urethane
  • Urethane
  • Urethane
  • Urethane
  • Seat dimensions:
  • Ø14×T1.5"
  • Ø14×T1.5"
  • Ø14×T1.5"
  • Ø14×T1.5"
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Foot Style
    Seat Height
    Availability
    Price
  • Lab chair, for bench height
    Black
    20" dia. adjustable chrome footring/black spoke; dual soft-wheel casters
    20.75 - 30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $682.12
    $682.12
    Prix promotionnel pour les nouveaux clients
    Each
       $682.12
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76308-804
    Model
    76308-804
    For use with
    Medium height benches (32.75 - 42")
    Seat/back color
    Black
    Seat/back material
    Urethane
    Seat dimensions
    Ø14×T1.5"
  • Lab chair, for bench height
    Black
    20" dia. adjustable chrome footring/black spoke; 2" nylon glides
    20.75 - 30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $855.34
    $855.34
    Each
       $855.34
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76308-806
    Model
    76308-806
    For use with
    Medium height benches (32.75 - 42")
    Seat/back color
    Black
    Seat/back material
    Urethane
    Seat dimensions
    Ø14×T1.5"
  • Lab chair, for desk height
    Black
    No footring; dual soft-wheel casters
    15 - 20"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $691.78
    $691.78
    Each
       $691.78
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76308-808
    Model
    76308-808
    For use with
    Low benches (27-32")
    Seat/back color
    Black
    Seat/back material
    Urethane
    Seat dimensions
    Ø14×T1.5"
  • Lab chair, for desk height
    Black
    No footring; 2" nylon glides
    15 - 20"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $685.60
    $685.60
    Each
       $685.60
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76308-810
    Model
    76308-810
    For use with
    Low benches (27-32")
    Seat/back color
    Black
    Seat/back material
    Urethane
    Seat dimensions
    Ø14×T1.5"