À propos de cet article
Competitive assay used for quantification of CysLT.
- Detect all CysLTs (LTC4, LTD4, and LTE4) in urine, tissue culture medium, and other sample matrices
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure CysLT levels down to 20 pg/ml
- Read: Colorimetric at 405 to 420 nm
Leukotrienes (LTs) are a group of acute inflammatory mediators derived from arachidonic acid via the 5-lipoxygenase (5-LO) pathway in leukocytes. LTC4, LTD4, and LTE4 are collectively referred to as cysteinyl leukotrienes (CysLTs).
LTC4 and LTD4 are potent mediators of asthma and hypersensitivity. They induce bronchoconstriction, increase microvascular permeability, and are vasoconstrictors of coronary arteries. The biological activity of LTE4 is much lower in most systems studied, but its presence reflects the prior existence of LTC4 and LTD4.
CysLT Express ELISA Kit is a competitive assay that can be used for measurement of all three primary CysLTs in urine, culture media, and other sample matrices. This sensitive assay takes about three hours to complete.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Assay Type:Competitive
- Developmental time:60 - 90 min
- Mid point:70 - 110 pg/ml (50% B/B0)
- Detection range:7.8 - 1000 pg/ml
- Storage temperature:−80 °C
- Sensitivity:20 pg/ml (80% B/B0)
- Incubation time:2 hr