Pneumatic Wheels and Axle Set for Single Gas Cylinder Hand Trucks, 10.5", Justrite®
76292-452
: Justrite
- Depth:10"
- Width:22"
- Height:10"
- Cat. no.:76292-452
- Weight:11 lbs.
Pneumatic Wheels and Axle Set for Single Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.
- Wheel and axle replacement set for single cylinder Justrite® hand trucks
- Wheels are 10.5×4"
- Axel size is 17.75×0.625"
: Includes two 10.5" pneumatic wheels with welded steel hubs and dual ball bearings, and plated axle with clips and washers.