wwPTFE (Water Wettable PTFE) Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Certificates
About this item
Next generation of universal hydrophilic PTFE membrane with maximum chemical compatibility when filtering both aqueous solutions and aggressive solvents.
- Hydrophilic
- Low protein binding
HPLC certified for low extractables assuring that the filter will not add artifacts or contamination to your analysis. Low protein binding of wwPTFE membrane provides high recovery of critical proteinaceous samples.
Frequently Bought Together