Ward's® Formaldehyde-Free Bullfrogs
470000-726
: Avantor
- Size:5 - 6"
- Injection Type:Double injected
- Cat. no.:470000-726
- Packaging:Pail of 10
- Supplier No.:WARD470000-726
Formaldehyde-free bullfrogs. Available in pails or vacuum packs.
- Specimens can be used comfortably over extended periods of time
To better serve the needs of all teaching environments and preferences, WARD’S® offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde is used in the fixing and preserving process for these specimens.
Instead, this solution used for preparation and binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’S® offers the largest selection of specimens prepared with this alternative process, including the widest range of sizes and injection styles.