AQ™ 96 Position Vials with Caps, MicroSolv
These vials are designed specifically for the Waters 96 position autosamplers so their performance is assured.
- Polyethylene caps with penetration point
These will not clog, bend or break the needles.
: Packed in AQ™ quality kit packs.
Frequently Bought Together