À propos de cet article
The durable, stainless steel-lined Protector Evidence Drying Cabinet provides a secure ventilated area to dry evidence such as clothing and bedding. The cabinet’s HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors. The spray nozzle allows the interior to be washed down for easy clean up after the cabinet has been properly disinfected. A built-in water pump aids clean up.
- Built-in blower with solid state speed control
- Epoxy-coated steel exterior
- Red, filter system alarm light to alert operator to low airflow conditions
- Safety glass door with magnetic seal that accommodates a user-supplied padlock or tamper-evident tag
- Water shut off valve
Activated organic vapor carbon filter removes nuisance odors from the air as it is exhausted from the cabinet to the room. Activated organic vapor carbon filter removes nuisance odors from the air as it is exhausted from the cabinet to the room. Inlet roughing prefilter traps dust and other large particles in the air
entering the cabinet. An exhaust roughing prefilter traps particulates in the exhaust air to prolong the life of the HEPA filter.
Built-in blower provides airflow through the cabinet interior to speed drying. Air passes through the door inlet prefilter, circulates up through the interior, passes through the exhaust prefilter, HEPA and carbon filters and is exhausted back to the room through the top of the cabinet.
Portable for easy installation. Cabinet dimensions accommodate passage through a standard door. Locking casters, 2" in diameter, simplify installation and allow the cabinet to be moved to storage or other location as needs change. A rear strap and bracket to secure the cabinet to the wall are provided. Storage area below the cabinet provides access to the spray nozzle and water shut off valve and can be used to house supplies.