$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Microbiology Test Systems
Endotoxin Tests
Control Standard Endotoxin (CSE), Charles River Endosafe MS
Control Standard Endotoxin (CSE), Charles River Endosafe MS
MSPP-E120
:
CHARLES RIVER ENDOSAFE MS
:
Control Standard Endotoxin (CSE), Charles River Endosafe MS
MSPP-E120
:
CHARLES RIVER ENDOSAFE MS
:
E120
:
Pack type:
Vial
Description:
Control Standard Endotoxin (CSE)
Size:
10 ng
Cat. no.:
MSPP-E120
Supplier No.:
E120