Distilling Adapters for Process Reactors, Chemglass
Supplier:   Chemglass
Distilling Adapters for Process Reactors, Chemglass
Supplier:  Chemglass
Certificates
Distilling Adapters for Process Reactors, Chemglass
Distilling Adapters for Process Reactors, Chemglass
2 Options Available Below
Product Details & Documents
Compact and simple distilling adapter for use with process reactors. Vapor flows upward freely and distillate is collected in to the small bowl below the joint and through the stopcock into a distilling receiver.
  • Lower stopcock is either a 6 mm or a 10 mm
  • Adapter is 6" wide overall with joints offset 95 mm center to center
Product Family Options
Adapter with Single Top Outer Joint
Informations sur le produit
  • Désignation
    ST Joint Size
    Stopcock Diameter
    Availability
    Price
  • Distilling Adapters with 3/4" Swagelock
    45/50
    10 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,040.00
    $1,040.00
    Each
       $1,040.00
    Supplier Number: CG-1235-52
Adapter with Two Top Outer Joints
Informations sur le produit
  • Désignation
    ST Joint Size
    Stopcock Diameter
    Availability
    Price
  • Distilling Adapters with 1/2" Swagelock
    24/40
    6 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $657.00
    $657.00
    Each
       $657.00
    Supplier Number: CG-1235-56