Pipette Filler, Safety Bulb, Hach
Catalog # CA14651-00
Supplier: Hach
Specifications
- Description:Pipette Filler, Safety Bulb
- Cat. no.:CA14651-00
Specifications
About this item
Unique cone adapter allows for universal pipette filling. Simply squeeze the rubber bulb to charge for filling, place pipette into cone adapter and insert into sample. Release the bulb to fill. Remove bulb and cover pipette end with finger for dispensing.
- Easy and safe
- Autoclavable
Delivery: Includes autoclavable elastic wristband.