ContactUsLinkComponent
Quantative Filter Papers, Pleated, Ashless and Hardened, Hach®
Quantative Filter Papers, Pleated, Ashless and Hardened, Hach®
Catalog # CA11024-814
Supplier:  Hach
Quantative Filter Papers, Pleated, Ashless and Hardened, Hach®
Catalog # CA11024-814
Supplier:  Hach
Supplier Number:  189457
Specifications
  • Ø:
    12.5 cm
  • Pore size:
    2 - 3 µm
  • Cat. no.:
    CA11024-814
Specifications
About this item
These filter papers are designed for use in various Hach Water Analysis Handbook Methods and other environmental reference methods. Optimized ashless hardened circles provide an abrasion-resistant smooth surface, which is ideal for gravimetric procedures utilizing Büchner funnels and high vacuum.
Pleated filters increase flow rates by doubling the effective filtration area of the same diameter filter circle compared to quadrant folded filters.