Simple Distillation Apparatus
Simple Distillation Apparatus
Catalog # 470310-686
Supplier: Wards
: CH0886
Jump to:
Product Details & Documents
This simple distillation apparatus is made up of boroscilicate glass.
: Apparatus includes round bottom flask (500 ml), still head with thermometer support, Liebig condensor (300 mm), receiver adapter and 3×joint clip (19/26).
Spécifications
- Désignation:Simple Distillation Apparatus