ContactUsLinkComponent
TPX Measuring Jugs
  470310-738
 :  Wards
 : CH0357K
undefined
TPX Measuring Jugs
  470310-738
 :  Wards
 :  CH0357I
 : CH0357K
Select an Option
100 ml
Select an Option
100 - 600 ml
Select an Option
600 ml
Select an Option
470310-738
These TPX jugs have a non-drip spout and excellent thumb grip on their handle making them perfect for every laboratory and classroom setting.
  • Fully autoclavable
  • Chemically resistant
  • Short form transparent jugs
Frequently Bought Together