Value ATR Top Plates, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
Value HATR trough/flat top-plates allow for simple sampling of liquids, solids, polymer films, powders, pastes, and gels.
Zinc Selenide crystal is designed for general purpose HATR sampling.
Caution: Avoid use with complexing agents, strong acids, bases, and oxidizers.
Frequently Bought Together